Magnetic stirring bars, double cross-head, PTFE
Supplier: COWIE
001.1135
COWI001.1135EA 10.1 SGD
COWI001.1135
Magnetic stirring bars, double cross-head, PTFE
Stirrers Magnetic Stirring Bars
Double cross-head has a greater turbulance when compared to single cross-head.