Magnetic stirring bars, PTFE, micro
Supplier: COWIE
001.830
COWI001.830EA 4.4 SGD
COWI001.830
Magnetic stirring bars, PTFE, micro
Stirrers Magnetic Stirring Bars
Magnetic stirrer bar with white PTFE coating, used for very small containers.