Stirring blades with square ends
Supplier: COWIE
002.052.1 002.076.1 002.090.1
COWI002.052.1EA 6.4 SGD
COWI002.052.1 COWI002.076.1 COWI002.090.1
Stirring blades with square ends
Stirrers Stirring Blades
PTFE. Blades fit PTFE stirring shaft; hole Ø 6,5 mm, 3,2 mm thick. Inert and non-scratch. The smooth surface prevents contamination and makes cleaning easier.
  • Excellent chemical resistance
  • Temperature resistant; suitable for use at high and low temperatures (−200 to +280 °C)