Stirring shafts, adjustable turbine
Supplier: COWIE
005.91075 005.912100 005.91275 005.916100 005.916150 005.90850 005.910100 005.90875
COWI005.91075EA 192 SGD
COWI005.91075 COWI005.912100 COWI005.91275 COWI005.916100 COWI005.916150 COWI005.90850 COWI005.910100 COWI005.90875
Stirring shafts, adjustable turbine
Stirrers Stirring Shafts
Adjustable turbine stirring shafts for use with plain glass, metal, or PTFE shafts.
Order Now
SPECIFICATIONS
Environmentally Preferable