Magnetic stirring bar, octagonal, PTFE
Supplier: COWIE
FT.8360
COWIFT.8360EA 0 SGD
COWIFT.8360
Magnetic stirring bar, octagonal, PTFE
Stirrers Magnetic Stirring Bars
Octahedral magnetic stirring bars.
  • Octahedral PTFE Stirrer Bar with champhered magnet core
  • 100% visual inspection using 4× magnification
Delivery information: Supplied with a free of charge certificate of analysis confirming the Job Number, material batch number, date of manufacture and inspection criteria.
Order Now
SPECIFICATIONS
Environmentally Preferable