Screw neck vials, 13-425 thread finish, Chromacol™
Supplier: CHROMACOL
5-SV 10-SV
CRMA5-SVEA 198 SGD
CRMA5-SV CRMA10-SV
Screw neck vials, 13-425 thread finish, Chromacol™
Vials
These vials are made from superior quality 1st hydrolytic class clear and amber glass (type 1).
Ordering information: Screw caps need to be ordered separately.
Order Now
SPECIFICATIONS
Environmentally Preferable