Storage vials, without caps, Chromacol™
Supplier: CHROMACOL
3.5-HRSV
CRMA3.5-HRSVEA 3530 SGD
CRMA3.5-HRSV
Storage vials, without caps, Chromacol™
Vials
These vials are manufactured from superior quality 33 expansion borosilicate clear (Type 1, Class A) glass and feature a 13-425 thread finish.