GC septa, Chromseal™ Chromacol™
Supplier: CHROMACOL
ECO-11 ECO-9.0 ECO-9.5 HT-11 HT-9.5 HT-SP LL-SP
CRMAECO-11EA 141 SGD
CRMAECO-11 CRMAECO-9.0 CRMAECO-9.5 CRMAHT-11 CRMAHT-9.5 CRMAHT-SP CRMALL-SP
GC septa, Chromseal™ Chromacol™
Septa
The CHROMSEAL 9001 autosampler septa are designed for use with GC autosamplers.
  • Provide better and cleaner chromatography
  • Offer a longer operating life
High performance septa (HT)- These red septa are designed to provide clean chromatography at a maximum operating temperature of 306 °C.
Long Life septa (LL)- These blue septa are designed for use with an autosampler where it is likely to be penetrated many times at a maximum operating temperature of 290 °C.
Economy septa (ECO)- These green septa are designed to provide low operating costs at aMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable