Stand-up pouches with zip closure, Lamizip
Supplier: DAKLAPACK
356 358 354 353 352 357 355 350
DAKL356EA 534 SGD
DAKL356 DAKL358 DAKL354 DAKL353 DAKL352 DAKL357 DAKL355 DAKL350
Stand-up pouches with zip closure, Lamizip
Bags Sample Bags
These stand-up pouches are suitable for products which need special requirements for packaging such as food, drink or pharmaceutical products. They can be used for the packaging of samples, substances and preparations.
  • Laminated foil with high barrier properties against oxygen and water vapour
  • Can be opened and closed innumerable times
  • Offer protection against the effects of UV radiation
  • Round bottom for increased stability
Order Now
SPECIFICATIONS
Environmentally Preferable