Laboratory glass cleaner, deconex® 12 BASIC
Supplier: BORER CHEMIE DECONEX
500325.00-K05W
DECO500325.00-K05WEA 510 SGD
DECO500325.00-K05W
Laboratory glass cleaner, deconex® 12 BASIC
Cleaning Agents Detergents and Surface Cleansers
Liquid, mildly alkaline cleaning concentrate for manual cleaning of laboratory glassware and reusable laboratory materials in ultrasonic and soaking baths.
  • Rinseable without residues and guarantees therefore consistent and error-free results
  • Perfect results even at low concentrations
  • High cleaning performance due to excellent swelling and dispersing capabilities
  • Working solution suitable for multiple use with a broad cleaning spectrum
  • Ideal as an adequate substitute for chromosulphuric acid, not toxic and environmentally friendly
  • EDTA-, NTA- and chlorine-free