Laboratory glass cleaner deconex®16 PLUS
Supplier: BORER CHEMIE DECONEX
509700.00-K05W
DECO509700.00-K05WEA 250 SGD
DECO509700.00-K05W
Laboratory glass cleaner deconex®16 PLUS
Cleaning Agents Detergents and Surface Cleansers
Liquid, pH-neutral cleaning concentrate for manual cleaning of laboratory glassware and reusable laboratory materials for use in ultrasonic and soaking baths.
  • Rinsable without residues and guarantees therefore consistent and error-free results
  • Perfect results even at low concentrations
  • Working solution suitable for multiple use
  • Good material compatibility, especially recommended for alkali and corrosion sensitive laboratory material, also suitable for aluminium
  • Phosphate-free, no release of phosphates into waste water
  • EDTA-, NTA- and chlorine-free, environmentally friendly