Pressure bulbs
Supplier: DEUTSCH NEUMANN
4200003
DENE4200003EA 19.2 SGD
DENE4200003
Pressure bulbs
Pipettes Pipette Bulbs
Natural rubber, chloroprene or PVC.
Used for filling automatic burettes, Pellet pattern (ball set with net) or atomisers (bulb).
Order Now
SPECIFICATIONS
Environmentally Preferable