Orsat Bladders (Scheibler Bladders) and Bulbs
Supplier: DEUTSCH NEUMANN
4400001
DENE4400001EA 24.7 SGD
DENE4400001
Orsat Bladders (Scheibler Bladders) and Bulbs
Bags Gas Bags
Natural rubber, transparent (Orsat bladders) or PVC, soft, maroon (inflation flask).
  • Orsat bladder (Scheibler bladder) for gas handling
  • Inflation bulb for a wide range of laboratory and industrial applications
  • Inflation bulb with suction and pressure valve