Metal-out bags, Statshield®
Supplier: DESCO
13050
DESC13050EA 96.5 SGD
DESC13050
Metal-out bags, Statshield®
Bags Storage Bags
Metal-Out ESD Shielding bags with low charging interior and exterior layers.
  • Aluminum metal outer layer of laminated film
  • Identified with ESD protective symbol
  • Lot coded
  • Polycarbonate compatible containing no amines or N-octanic acid
  • Testable to industry standards
  • Antistatic, low charging, reclosable zipper
40% light transmission allows for identification of bag contents. Integral antistatic, low tribo charging properties bag contents will not electrostatically charge during movement. Integral static dissipative properties not topically treated, notMore Product Information