ORCA Response Temperature-Controlled Packaging Solution
Supplier: DGP INTELSIUS
ORS200DI ORS223 ORSBAG2L
DGINORS200DIEA 0 SGD
DGINORS200DI DGINORS223 DGINORSBAG2L
ORCA Response Temperature-Controlled Packaging Solution
Mailers and Shipping Containers
ORCA Response is a multi-use, handheld, temperature-controlled packaging solution for transporting smaller payloads of red blood cells, platelets, pharmaceutical products and be easily carried and stored, making it the ideal solution for taking personal medicines on long journeys, including suitability as hand luggage on aeroplanes.
  • Easy to assemble
  • Utilises Phase Change Material (PCM) and Vacuum Insulation Panel (VIP) technologies
  • Universal pack out, Simple conditioning and assembly
  • 3 PCM temperature ranges: 2...8 ˚C, 15...25 ˚C, –20 ˚C
  • Qualified for
More Product Information