Intelsius® PharmaTherm Temperature Controlled Packaging
Supplier: DGP INTELSIUS
PHT019 PHT020 PHT021 PHT001
DGINPHT019EA 232 SGD
DGINPHT019 DGINPHT020 DGINPHT021 DGINPHT001
Intelsius® PharmaTherm Temperature Controlled Packaging
Mailers and Shipping Containers
Utilises pharmachill cool packs are hard wearing and durable re-usable coolant, reducing costs and landfill waste.
2 to 8 °C systems qualified against the Intelsius high performance quad profile with stress peaks and overlapping mid season temperatures, the PharmaTherm offers robust performance for all season shipping and peace of mind.
15 to 25 °C systems qualified against Intelsius controlled room temperatures profiles for ambient shipments. GDP compliant systems.