Chart for vaccine chart recorder, 4"
Supplier: DICKSON
C204
DICKC204EA 0 SGD
DICKC204
Chart for vaccine chart recorder, 4"
Chart Recorders Charts for Recorders
The vaccine chart recorder can be placed right inside the refrigerator or freezer along with the vaccines.
  • Easy-to-read special vaccine chart with circular bands highlight the acceptable vaccine ranges of 2 to 8 °C
  • LCD display shows current temperature for quick reference