Temperature chart recorders, disposable
Supplier: DICKSON
D302
DICKD302EA 0 SGD
DICKD302
Temperature chart recorders, disposable
Chart Recorders Recorders
These are single use logger accurately records temperatures from start to finish of transport and documents adherence to critical storage environments.
  • Failsafe start-up and pull out start tab
  • Tamper-proof crimped seal
  • Shock and vibration protection
Delivery information: Supplied with a shipment/job sheet and a mounting tape.
Order Now
SPECIFICATIONS
Environmentally Preferable