Voltage chart recorder
Supplier: DICKSON
ET855
DICKET855EA 0 SGD
DICKET855
Voltage chart recorder
Chart Recorders Recorders
This chart recorder is designed for a wide variety of applications. It features high/low audible alarm, user calibration, large display, user selectable output ranges and recording times, and two input channels.
  • The high/low alarm displays minimum/maximum readings with no controller/relay functions
  • Two screw terminals on the back of the unit allow for easy connection of input transmitter
  • Free-standing or wall-mountable
Delivery information: Supplied with two pens (red and blue), a 240 VAC adapter and a nine volt battery.