Temperature Loggers
Supplier: DICKSON
SM420
DICKSM420EA 0 SGD
DICKSM420
Temperature Loggers
Data Loggers
These temperature loggers are designed for monitoring, recording and evaluating temperature of critical equipment, computers, HVAC/R systems, incubators, freezers, autoclaves, baths and cleanrooms. They have a high/low audible alarms.
  • Large four-digit LCD features
  • Flash memory option for easy data transfer
  • USB enabled fast downloading
  • Min/max display
  • Temperature model and remote probe model
  • Super high speed USB 2.0 connectivity
  • Download data faster when compared to using standard serial port
A memory card reader kit and removeable Flash memory storageMore Product Information