Counting chamber with grid
Supplier: DIVERSIFIED BIOTECH
GRID-1000
DIVEGRID-1000EA 132 SGD
DIVEGRID-1000
Counting chamber with grid
Slides Counting Chambers
Disposable polyester with acrylic adhesive grid stickers for micro or semi-micro objects.
  • Inert and stable under most conditions
  • Compatible with different kinds of microscopes
  • Separate coverslips can be placed over each sticker
These slide grids are either used with dry or suspended specimens, such as cells held in buffer, under cover slips.
Order Now
SPECIFICATIONS
Environmentally Preferable