Lab markers, Teeny Writers
Supplier: DIVERSIFIED BIOTECH
TNY-TWIN
DIVETNY-TWINEA 28.7 SGD
DIVETNY-TWIN
Lab markers, Teeny Writers
Pens Lab Markers
A retractable Sharpie® pen with an extra fine tip. Recommended for writing on all our labels for samples that will be stored at room temperature and not exposed to chemicals and/or caustic agents.
Order Now
SPECIFICATIONS
Environmentally Preferable