Bellows pumps, accuro®
Supplier: Dräger
6400000
DRAE6400000EA 2310 SGD
DRAE6400000
Bellows pumps, accuro®
Gas Detection Systems
Ergonomic pump for use with short-term tubes.
  • Stroke counter
  • Final pump stroke indicator
  • Made from solid synthetic material