RD40 Respirator filters for X-plore® masks
Supplier: Dräger
6738855
DRAE6738855EA 82.5 SGD
DRAE6738855
RD40 Respirator filters for X-plore® masks
Respirators Reusable Respirators Respirator Filters
The X-plore® Rd40 filter with aluminium housing is suitable for half and full face masks with connectors conforming to EN 148-1.
Certifications: EN 141:2000, EN 14387, EN 143:2000, EN 371