Gas detection tube for hydrogen sulphide
Supplier: Dräger
8101831
DRAE8101831EA 405 SGD
DRAE8101831
Gas detection tube for hydrogen sulphide
Gas Tubes
To measure the 'spot concentrations' of hydrogen sulphide for use with an Accuro® pump. Uses include determination and measurement of concentration peaks, measurement of personal exposure in the inhalation area, detection of leaks in, for example, pipelines or gas installations, and air analysis in sewers, shafts and confined spaces.
One stroke is equivalent to a volume of 100 ±5 ml of gas passing through the tube.
The standard measuring range is calibrated at 20 °C and 1013 hPa.
Operating conditions:
0,2/a: 10 to 30 °C ; 3 to 15 mg/l H₂O absolute humidity
0,2/b: 15 to 40 °C ; Up to 2More Product Information