Gas detection tubes for water vapour
Supplier: Dräger
8103061
DRAE8103061EA 388 SGD
DRAE8103061
Gas detection tubes for water vapour
Gas Tubes
To measure the 'spot concentrations' of water vapour for use with an Accuro® pump.
Uses include determination and measurement of concentration peaks, measurement of personal exposure in the inhalation area, detection of leaks in pipelines or gas installations.
One stroke is equivalent to a volume of 100 ±5 ml of gas passing through the tube.
The standard measuring range is calibrated at 20 °C and 1013 hPa.