Microtest manifolds
Supplier: DRUMMOND
3-000-096
DRUM3-000-096EA 826 SGD
DRUM3-000-096
Microtest manifolds
Manifolds
Stainless steel/PTFE manifolds allow multiple test positions to be fed, rinsed, or aspirated quickly and easily.
  • Autoclavable
  • Use for both sample delivery and aspiration operations
  • Supplied complete with cleaning wire for easy cleaning
  • Positive Luer lock connection