Safety cabinets, type 90, with wing door technology, CLASSIC line standard and CLASSIC line pro
Supplier: DUPERTHAL
29-201267-030
DUEH29-201267-030EA 8970 SGD
DUEH29-201267-030
Safety cabinets, type 90, with wing door technology, CLASSIC line standard and CLASSIC line pro
Furniture Storage Cabinets
Safety cabinets with storage shelves (CLASSIC line standard) or pull-out shelves (CLASSIC line pro) for the storage of hazardous substances in working spaces. Complete carcass, interior fittings and wing doors are made of powder-coated sheet steel.
  • Doors with standard integrated arrest system, meaning that containers can be placed into and removed from the cabinet with both hands
  • Removable base and adjustable feet for easy alignment and transport with a hand pallet truck.
  • The base is set back to prevent hitting one’s feet
  • Ventilation at every cabinet
More Product Information