Gowns, Tyvek® IsoClean®, model IC 701
Supplier: DuPont
IC701SWHOO
DUPOIC701SWHOOEA 359 SGD
DUPOIC701SWHOO
Gowns, Tyvek® IsoClean®, model IC 701
Disposable Garments
These gowns are made of Tyvek® which is a fabric composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort.
  • Ultra-low linting fabric
  • Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam
  • Secure, knit cuffs for comfort and fit
  • Bound neck for lower particle shedding with ties for an adjustable fit
  • Bound ties at the waist originate at the front and tie in the back
Tyvek® is permeable to both air and waterMore Product Information