Cleanroom face masks, Tyvek™ Sierra™
Supplier: DuPont
ML7360WH000S
DUPOML7360WH000SEA 545 SGD
DUPOML7360WH000S
Cleanroom face masks, Tyvek™ Sierra™
Masks Cleanroom Masks
Cleanroom face masks provide high breathability with exceptional filtration efficiency.
  • Pleated rayon outer facing
  • Metal nose piece
  • 23 cm bound Tyvek® ties