Chemical resistant boot covers, Tychem® F, model POBA
Supplier: DuPont
TYFPOBASGY00
DUPOTYFPOBASGY00EA 706 SGD
DUPOTYFPOBASGY00
Chemical resistant boot covers, Tychem® F, model POBA
Disposable Garments
Specially designed for use with Tychem® apparel, these boot covers can help offer enhanced protection for body parts that are more exposed to hazardous substances.
  • Knee-length boot covers with slip-retardant sole and fixation ties
  • Sole is partially stitched: splash-proof, not fully liquid tight
  • Stitched and over-taped seams with barrier tape for protection and strength
  • Anti-static treatment (EN 1149-5) inside only
  • Conform to EN 14126 (barrier to infective agents)
Tychem® F boot covers are made from a lightweight and durable fabric consisting of aMore Product Information