Core thermometer, TDC 150
Supplier: EBRO
1340-1611
EBRO1340-1611EA 140 SGD
EBRO1340-1611
Core thermometer, TDC 150
Thermometers Digital Thermometers
Core thermometer works with fast response time.
  • Fixed probe, pointed
  • Robust and impact-resistant
  • Replaceable battery
  • Switchable between °C/°F