Disposal bins for serological pipettes, Bio-bin®
Supplier: ECONIX LTD
BEXBBPIP6TS
ECONBEXBBPIP6TSEA 431 SGD
ECONBEXBBPIP6TS
Disposal bins for serological pipettes, Bio-bin®
Boxes Waste Disposal Boxes
The 6 L Bio-bin® has been specifically designed to safely contain serological pipettes in labs.
  • Tall and thin: Best used with a stand
  • Fold-flat space efficient design, stackable pre- and post-use
  • Absorbent Bio-matt™ in the base of every bin can absorb 100 ml of fluid
  • Built-in lid and handles
  • Temporary and permanent seals
  • Fully autoclavable
Order Now
SPECIFICATIONS
Environmentally Preferable