Organic reference standard, Betamethasone acetate, Ph.Eur., RS
Supplier: EDQM
B1030000
EDQMB1030000EA 441 SGD
EDQMB1030000
Organic reference standard, Betamethasone acetate, Ph.Eur., RS
Standards Organic Standards
  • In Monographs 0548 and 0975
  • Solubility in water: less than 0,1 g/l