Organic reference standards, betaxolos, Ph. Eur.

Supplier: EDQM

B1103000
EDQMB1103000EA 441 SGD
EDQMB1103000
Organic reference standards, betaxolos, Ph. Eur.
Standards Organic Standards

Order Now

SPECIFICATIONS

Description Betaxolol hydrochloride
Environmentally Preferable
CAS No. 63659-19-8
Formula C₁₈H₃₀ClNO₃

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR