tert-Butyl-4-methoxyphenol (mixture of isomers)

Supplier: EDQM

Synonyms: tert-Butyl-4-methoxyphenol, BHA, tert-Butyl-4-hydroxyanisole, Butylhydroxyanisole, 2(3)-tert-Butyl-4-methoxyphenol, Butylated hydroxyanisole

B1212000
EDQMB1212000EA 419 SGD
EDQMB1212000
tert-Butyl-4-methoxyphenol (mixture of isomers)
tert-Butyl-4-methoxyphenol (mixture of isomers)
Formula: C₁₁H₁₆O₂
Boiling Pt: 264 °C (1013 hPa)
Melting Pt: 48…55 °C
Density: 1,05 g/cm³ (20 °C)
Flash Pt: 156 °C
Storage Temperature: Ambient
MDL Number: MFCD01779059
CAS Number: 25013-16-5
EINECS: 246-563-8
Merck Index: 12,01582

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR