Carbenicillin disodium salt

Supplier: EDQM
Danger

C0455000
EDQMC0455000EA 441 SGD
EDQMC0455000
Carbenicillin disodium salt
Carbenicillin disodium salt
Formula: C₁₇H₁₆N₂O₆S·2Na
Storage Temperature: Refrigerator
MDL Number: MFCD00077683
CAS Number: 4800-94-6
EINECS: 225-360-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR