Organic reference standard, clotrimazole, Ph.Eur., RS
Supplier: EDQM
C2430000
EDQMC2430000EA 441 SGD
EDQMC2430000
Organic reference standard, clotrimazole, Ph.Eur., RS
Standards Organic Standards
  • In Monograph 0757
  • Soluble in/miscible with acetone, chloroform, ether, methanol