Human albumin (from serum)

Supplier: EDQM

H0900000
EDQMH0900000EA 441 SGD
EDQMH0900000
Human albumin (from serum)
Proteins and Peptides

Order Now

SPECIFICATIONS

Protein/peptide name Albumin
Environmentally Preferable
Protein synonyms Human Serum Albumin|HSA|serum albumin|HAS|serum of albumin
Species Human
Source Serum
CAS No. 70024-90-7
Storage conditions Refrigerator

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR