Pilocarpine hydrochloride

Supplier: EDQM

Synonyms: (+)-Pilocarpine hydrochloride

P1650000
EDQMP1650000EA 441 SGD
EDQMP1650000
Pilocarpine hydrochloride
Pilocarpine hydrochloride
Formula: C₁₁H₁₇ClN₂O₂
MDL Number: MFCD00012722
CAS Number: 54-71-7
EINECS: 200-212-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR