Organic reference standards, prednisolones, Ph. Eur., RS

Supplier: EDQM

P2804000 P2700000 Y0000582
EDQMP2804000EA 441 SGD
EDQMP2804000 EDQMP2700000 EDQMY0000582
Organic reference standards, prednisolones, Ph. Eur., RS
Standards Organic Standards

Order Now

SPECIFICATIONS

Description Prednisolone hexanoate Prednisolone Prednisolone acetate for peak identification
Environmentally Preferable
CAS No. 7681-14-3 50-24-8 52-21-1
Formula C₂₇H₃₈O₆ C₂₁H₂₈O₅ C₂₃H₃₀O₆

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR