Polymyxin B sulphate

Supplier: EDQM

Y0000355
EDQMY0000355EA 441 SGD
EDQMY0000355
Polymyxin B sulphate
Polymyxin B sulphate
Formula: C₅₅H₉₆N₁₆O₁₃·2H₂SO₄
MDL Number: MFCD00131991
CAS Number: 1405-20-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR