Digoxin

Supplier: EDQM

Y0000554
EDQMY0000554EA 419 SGD
EDQMY0000554
Digoxin
Digoxin
Formula: C₄₁H₆₄O₁₄
MW: 780.95 g/mol
MDL Number: MFCD00003674
CAS Number: 20830-75-5
EINECS: 244-068-1
UN: 2811
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR