Capsaicin

Supplier: EDQM

Synonyms: 8-Methyl-N-vanillyl-6-none-amid

Y0000671
EDQMY0000671EA 441 SGD
EDQMY0000671
Capsaicin
Capsaicin
Formula: C₁₈H₂₇NO₃
MW: 305.42 g/mol
Boiling Pt: 210…220 °C (0.01 mmHg)
Melting Pt: 66.1
MDL Number: MFCD00017259
CAS Number: 404-86-4
EINECS: 206-969-8
UN: 2811
ADR: 6.1 III
Merck Index: 12,01811

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR