Organic reference standards, levetiracetams, Ph. Eur.

Supplier: EDQM

Y0001253
EDQMY0001253EA 419 SGD
EDQMY0001253
Organic reference standards, levetiracetams, Ph. Eur.
Standards Organic Standards

Order Now

SPECIFICATIONS

Description Levetiracetam
Environmentally Preferable
CAS No. 102767-28-2
Formula C₈H₁₄N₂O₂

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR