Bags, Amber
Supplier: ELKAY
FAM30406 FAM31212
ELKAFAM30406EA 139 SGD
ELKAFAM30406 ELKAFAM31212
Bags, Amber
Bags Storage Bags
Amber bags prevent their contents from being degraded or damaged by UV light waves.
  • Open-ended bags can be sealed with a heat sealer or taped shut
  • Seal top bags can be accessed multiple times
They are a choice for packaging and transporting medications and other UV light-sensitive products.
Order Now
SPECIFICATIONS
Environmentally Preferable