Modular Basket System
Supplier: ELMA SCHMIDBAUER
1108916
ELMA1108916EA 144 SGD
ELMA1108916
Modular Basket System
Ultrasonic Baths
Modular basket system for Elmasonic P, Elmasonic Select and EASY series ultrasonic baths.
  • Stainless steel with plastic handles (10 to 120 and 180 to 300) or Rilsan coating (150)
  • Ergonomic handles
  • Autoclavable
  • Bayonet holes in the bottom for mounting holders (e.g. Elma Pin, silicone holder bottom comb)
  • Square openings in the side walls for mounting silicone holders
Hole diameter bottom (mm): 11×15
Hole diameter sides (mm): 10×10