Cleaning agents, elma tec clean
Supplier: ELMA SCHMIDBAUER
8000152
ELMA8000152EA 114 SGD
ELMA8000152
Cleaning agents, elma tec clean
Cleaning Agents Detergents and Surface Cleansers
Powerful cleaners for ultrasonic baths suitable for many industrial cleaning tasks
  • prolongs the service life of the ultrasonic unit
  • Highly efficient concentrate
  • Long shelf life
Order Now
SPECIFICATIONS
Environmentally Preferable