Pipette tips, epT.I.P.S.® reloads (IVD marked)
Supplier: EPPENDORF
0030073.444
EPPE0030073.444EA 141 SGD
EPPE0030073.444
Pipette tips, epT.I.P.S.® reloads (IVD marked)
Pipette Tips
A compact system for refilling epT.I.P.S.® available as one filled box of tips; a set comprising one box and five trays of tips; and reloads of ten tip trays. Components are 100% recyclable.
  • Optimised system for multi-channel pipettes
  • Reloads available in a PCR clean version: Free of human DNA, DNase, RNase and PCR inhibitors
Designed and constructed for low-contamination transfer of liquids, in particular for samples from the human body and for reagents within the scope of an in-vitro diagnostic application in order to allow the in-vitro diagnosticMore Product Information