Dispenser tips, Combitips advanced® and ViscoTip®
Supplier: EPPENDORF
0030089.502
EPPE0030089.502EA 0 SGD
EPPE0030089.502
Dispenser tips, Combitips advanced® and ViscoTip®
Pipette Tips
Combitips advanced® function according to the positive displacement principle, as a result they always dispense the correct volume independently of the density of the liquid and its flow characteristics (e.g., increased vapour pressure or increased viscosity). They are ideal for use with all Eppendorf multipettes.
  • Ergonomic design with unique funnel shape prevents damage to gloves ensuring comfortable handling
  • Individually colour coded for quick identification
  • Elongated tips (for 2,5; 5 and 10 ml) allow complete emptying of all common tubes preventing
More Product Information